ContactUsLinkComponent
Dish Evaporating,150ml,VWR PTFE Tall Form w Spout,Ht:43xDia:78mm,E
Catalog # COWI006.150
Supplier:  COWIE
Dish Evaporating,150ml,VWR PTFE Tall Form w Spout,Ht:43xDia:78mm,E
Catalog # COWI006.150
Supplier:  COWIE