2,6-Diethylaniline 98%
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
2,6-Diethylaniline 98%
Catalog # ACRO165222500
Supplier: Thermo Fisher Scientific
CAS Number: 579-66-8
Jump to:
Product Details & Documents
- CAS Number:579-66-8
- Formula:C₁₀H₁₅N
- MDL Number:MFCD00007753
- Molecular Weight:149.24 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:3…4 °C
- Density:0.9 g/cm³
- Flash Pt:123 °C
- Storage Temperature:Ambient
- EINECS:209-445-7
Documents
ACRO16522
SDS
Specifications
Specification Test Results
- Appearance:Clear yellow to red-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5446 to 1.5466 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:250 mL



