Order Entry
Export
Orders LinkContactUsLinkComponent
Acrylamide 98.5%, extra pure

Acrylamide 98.5%, extra pure

Plastic bottle, 2,5 kg
Catalog # ACRO164830025
Toxic
Health
Acrylamide 98.5%, extra pure
Catalog # ACRO164830025
CAS Number:  79-06-1
Acrylamide 98.5%, extra pure
Acrylamide 98.5%, extra pure

Plastic bottle, 2,5 kg

CAS Number:  79-06-1
Price
132,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:125 °C (25 mmHg)
  • Melting Pt:82…86 °C
  • Density:1.122 g/cm³
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    82 to 86 °C
  • Water:
    ≤2.0 %
  • GC:
    ≥98.0 %
  • Color of solution:
    (10 % in water) Clear and colorless

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 kg

Specifications