Order Entry
Export
Orders LinkContactUsLinkComponent
Pyrogallol 99%

Pyrogallol 99%

Plastic bottle, 100 g
Catalog # ACRO164561000
Toxic
Health
Pyrogallol 99%
Catalog # ACRO164561000
CAS Number:  87-66-1
Pyrogallol 99%
Pyrogallol 99%

Plastic bottle, 100 g

CAS Number:  87-66-1
Price
94,10 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
  • CAS Number:87-66-1
  • Formula:C₆H₆O₃
  • MDL Number:MFCD00002192
  • Molecular Weight:126.11 g/mol
  • Merck Index:13,08090
  • Boiling Pt:309 °C (1013 hPa)
  • Melting Pt:131…135 °C
  • Density:1,453 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:201-762-9
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    131 to 135 °C
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥98.5 %
  • Arsenic (As):
    ≤0.5 ppm
  • Calcium (Ca):
    ≤20 ppm
  • Iron (Fe):
    ≤5 ppm
  • Magnesium (Mg):
    ≤5 ppm
  • Potassium (K):
    ≤20 ppm
  • Heavy metals (as Pb):
    ≤5 ppm
  • Nitrogen compounds (as N):
    ≤5 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications