2,5-Dimethyl-2,5-hexanediol 97%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
2,5-Dimethyl-2,5-hexanediol 97%
Catalog # ACRO162502500
Supplier: Thermo Fisher Scientific
CAS Number: 110-03-2
Jump to:
Product Details & Documents
Synonyms: 1,1,4,4-Tetramethyl-1,4-butanediol
- CAS Number:110-03-2
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00004473
- Molecular Weight:146.23 g/mol
- Boiling Pt:214 °C (1013 hPa)
- Melting Pt:88…91 °C
- Density:0,898 g/cm³ (20 °C)
- Flash Pt:126 °C
- Storage Temperature:Ambient
- EINECS:203-731-5
Specifications
Specification Test Results
- Appearance:White Flakes or pellets or prills
- Infrared spectrum:Conforms
- Melting point:85 to 91 °C
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



