Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mesitylene 97%, pure

Glass bottle, 250 mL
Catalog # ACRO161322500
Irritant
Environment
Flammable
Health
Mesitylene 97%, pure
Catalog # ACRO161322500
CAS Number:  108-67-8
Mesitylene 97%, pure
Mesitylene 97%, pure

Glass bottle, 250 mL

CAS Number:  108-67-8
Price
46,50 €
Log in for availability
1 * 250 mL
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:163…166 °C (760 mmHg)
  • Melting Pt:–45 °C
  • Density:0.8675 g/cm³
  • Flash Pt:44 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4980 to 1.5000 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications