Order Entry
Export
Orders LinkContactUsLinkComponent
4-Methoxybenzonitrile 99%

4-Methoxybenzonitrile 99%

Glass bottle for solids, 25 g
Catalog # ACRO158050250
Irritant
4-Methoxybenzonitrile 99%
Catalog # ACRO158050250
CAS Number:  874-90-8
4-Methoxybenzonitrile 99%
4-Methoxybenzonitrile 99%

Glass bottle for solids, 25 g

CAS Number:  874-90-8
Price
81,90 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
  • CAS Number:874-90-8
  • Formula:C₈H₇NO
  • MDL Number:MFCD00001818
  • Molecular Weight:133.15 g/mol
  • Boiling Pt:256…257 °C (765 mmHg)
  • Melting Pt:57…60 °C
  • Storage Temperature:Ambient
  • EINECS:212-871-6
  • UN:3439
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    57 to 60 °C
  • GC:
    ≥98.5 %
  • UV:
    at 205 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Acidity/alkalinity:
    ≤0.0001 meq/g

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications