Order Entry
Export
Orders LinkContactUsLinkComponent
Cell culture inserts, Netwell™

Cell culture inserts, Netwell™

Membrane Ø 24 mm, mesh size 74 μm, 6/plate
Catalog # 734-1588
   Low Manufacturing Impact
Supplier:   Corning
Cell culture inserts, Netwell™
Catalog # 734-1588
Supplier:  Corning
   Low Manufacturing Impact
Cell culture inserts, Netwell™
Cell culture inserts, Netwell™

Membrane Ø 24 mm, mesh size 74 μm, 6/plate

Price
461,00 €
Log in for availability
1 * 48 items
Product Details & Documents

Polyester mesh bottoms attached to PS rings or housing. Netwell™ inserts are used as tissue carriers, supports and strainers of small organs, tissue slices or explants at the air-media interface.

  • Handy carrier for immunocytochemical staining of tissue slices
  • Provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
  • Available in two mesh sizes and diameters
  • Supplied sterile and pre-loaded in 6- or 12-well plates
  • 24 mm Netwell inserts fit in Corning® 50 ml plastic centrifuge tubes

Polystyrene reagent trays are available in black for improved visibility of tissue sections, or white for colorimetric reaction contrast. Specially designed carriers and handles permit the simultaneous processing of up to 12 samples per carrier. Each carrier kit contains eight carriers and eight handles.

Specifications
  • Packed:
    6/plate
  • Description:
    Membrane Ø 24 mm, mesh size 74 μm
  • Cat. No.:
    734-1588
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Netwell™ carrier kit, 24 mm
    Netwell™ carrier kit, 24 mm
    734-1603
    Log in for availability
    1 * 8 items - 175,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Netwell™ carrier kit, 15 mm
    Netwell™ carrier kit, 15 mm
    734-1602
    Log in for availability
    1 * 8 items - 180,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Netwell™ reagent tray, white
    Netwell™ reagent tray, white
    734-1601
    Log in for availability
    1 * 200 items - 267,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Netwell™ reagent tray, black
    Netwell™ reagent tray, black
    734-1600
    Log in for availability
    1 * 200 items - 267,00 €