Order Entry
Export
Orders LinkContactUsLinkComponent
96-well plates, polypropylene, without lids

96-well plates, polypropylene, without lids

Storage plate, 96-well, black, round well, 360 µl, -, 25/bag
Catalog # 734-1782
   Low Manufacturing Impact
Supplier:   Corning
96-well plates, polypropylene, without lids
Catalog # 734-1782
Supplier:  Corning
   Low Manufacturing Impact
96-well plates, polypropylene, without lids
96-well plates, polypropylene, without lids

Storage plate, 96-well, black, round well, 360 µl, -, 25/bag

Price
542,00 €
Log in for availability
1 * 100 items
Product Details & Documents

Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.

  • Flat, round, or V-shaped well bottom
  • Features uniform skirt heights for greater robotic gripping surface
  • Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
  • RNase-/DNase-free
  • Available sterile or non sterile

 : Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.

Specifications
  • Packed:
    25/bag
  • Sterile:
    -
  • Description:
    Storage plate, 96-well, black, round well
  • Seq:
    04
  • Well volume:
    360 µl
  • Cat. No.:
    734-1782
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact