96-well plates, polypropylene, without lids
Storage plate, 96-well, white, round well, 360 µl, -, 25/bag
Supplier: Corning
96-well plates, polypropylene, without lids
Catalog # 734-1781
Supplier: Corning
Jump to:
Product Details & Documents
Corning polypropylene microplates offer both small volume and large volume (blocks) well formats to meet assay and storage requirements.
- Flat, round, or V-shaped well bottom
- Features uniform skirt heights for greater robotic gripping surface
- Solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol)
- RNase-/DNase-free
- Available sterile or non sterile
: Plates are packaged 25 per pack, 4 packs per case. Nonsterile 450 µL plate is packaged 10 plates per bag and 5 bags per case.
Specifications
- Packed:25/bag
- Sterile:-
- Description:Storage plate, 96-well, white, round well
- Seq:03
- Well volume:360 µl
- Cat. No.:734-1781
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact



