2-Mercaptopyrimidine 98%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
2-Mercaptopyrimidine 98%
Catalog # ACRO156740500
Supplier: Thermo Fisher Scientific
CAS Number: 1450-85-7
Jump to:
Product Details & Documents
Synonyms: 2-Thiopyrimidine
- CAS Number:1450-85-7
- Formula:C₄H₄N₂S
- MDL Number:MFCD00006073
- Molecular Weight:112.16 g/mol
- Melting Pt:230 °C
- Storage Temperature:Ambient
- EINECS:215-917-3
Documents
ACRO15674
SDS
Specifications
Specification Test Results
- Appearance:Yellow Powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %
- Phosphate (PO4):≤5 ppm
- Ammonium (NH4):≤5 ppm
- Calcium (Ca):≤0.002 %
- Iron (Fe):≤5 ppm
- Magnesium (Mg):≤0.001 %
- Sodium (Na):≤0.03 %
- Heavy metals (as Pb):≤5 ppm
- Sulfur compounds (as SO4):≤0.003 %
- Insoluble matter:≤0.01 %
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



