2,3-Diaminopyridine 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2,3-Diaminopyridine 98%
Catalog # ACRO155020250
Supplier: Thermo Fisher Scientific
CAS Number: 452-58-4
Jump to:
Product Details & Documents
Synonyms: Pyridine-2,3-diyldiamine
- CAS Number:452-58-4
- Formula:C₅H₇N₃
- MDL Number:MFCD00006319
- Molecular Weight:109.13 g/mol
- Boiling Pt:148…150 °C (5 mmHg)
- Melting Pt:114…116 °C
- Flash Pt:205 °C
- Storage Temperature:Refrigerator
- EINECS:207-200-9
- UN:2811
- ADR:6.1,II
Specifications
Specification Test Results
- Appearance:Grey to dark brown Crystalline powder or needles (May darken on storage)
- Infrared spectrum:Conforms
- GC:≥97.5 %
- Titration with HClO4:97.5 to 102.5 %
- NMR (1H-NMR):Consistent with structure
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



