Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexene 99% stabilised, pure

Cyclohexene 99% stabilised, pure

Glass bottle, 2,5 L
Catalog # ACRO154840025
Irritant
Health
Flammable
Environment
Cyclohexene 99% stabilised, pure
Catalog # ACRO154840025
CAS Number:  110-83-8
Cyclohexene 99% stabilised, pure
Cyclohexene 99% stabilised, pure

Glass bottle, 2,5 L

CAS Number:  110-83-8
Price
127,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
  • CAS Number:110-83-8
  • Formula:C₆H₁₀
  • MDL Number:MFCD00001539
  • Molecular Weight:82.15 g/mol
  • Merck Index:13,02756
  • Boiling Pt:83 °C (760 mmHg)
  • Melting Pt:–104 °C
  • Density:0.81 g/cm³
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • EINECS:203-807-8
  • UN:2256
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4450 to 1.4480 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Stabilizer:
    Approx. 100 ppm 2-tert-Butyl-4-methylphenol

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications