3,3-Dimethylpentane 99%
Glass bottle, 1 g
- :562-49-2
- :C₇H₁₆
- :MFCD00009322
- :100.2 g/mol
- :86 °C (760 mmHg)
- :–135 °C
- :0.69 g/cm³
- :–6 °C
- :Ambient
- :209-230-8
- :3,II
- :Density: 0.6936 g/cm³ (20 °C)
- :1206
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3900 to 1.3920 (20°C, 589 nm)
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:1 g



