1,3-Dichlorobenzene 98%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
1,3-Dichlorobenzene 98%
Catalog # ACRO151180010
Supplier: Thermo Fisher Scientific
CAS Number: 541-73-1
Jump to:
Product Details & Documents
- CAS Number:541-73-1
- Formula:C₆H₄Cl₂
- MDL Number:MFCD00000573
- Molecular Weight:147 g/mol
- Merck Index:13,03080
- Boiling Pt:173 °C (1013 hPa)
- Melting Pt:–24 °C
- Density:1.28 g/cm³
- Flash Pt:67 °C
- Storage Temperature:Ambient
- EINECS:208-792-1
- UN:3082
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5450 to 1.5470 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:1 L



