Acetanilide 99+%
Plastic bottle, 500 g
: N-Phenylacetamide
- :103-84-4
- :C₈H₉NO
- :MFCD00008674
- :135.17 g/mol
- :13,00051
- :304 °C (760 mmHg)
- :111…115 °C
- :1,22 g/cm³ (20 °C)
- :173 °C
- :Ambient
- :203-150-7
- Appearance:White to beige to grey to light brown Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:111 to 115 °C
- GC:≥99.0 %
- Pack type:Plastic bottle
- Pk:500 g



