Order Entry
Export
Orders LinkContactUsLinkComponent
Acetanilide 99+%

Acetanilide 99+%

Plastic bottle, 250 g
Catalog # ACRO150812500
Irritant
Acetanilide 99+%
Catalog # ACRO150812500
CAS Number:  103-84-4
Acetanilide 99+%
Acetanilide 99+%

Plastic bottle, 250 g

CAS Number:  103-84-4
Price
39,10 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  N-Phenylacetamide
  • CAS Number:103-84-4
  • Formula:C₈H₉NO
  • MDL Number:MFCD00008674
  • Molecular Weight:135.17 g/mol
  • Merck Index:13,00051
  • Boiling Pt:304 °C (760 mmHg)
  • Melting Pt:111…115 °C
  • Density:1,22 g/cm³ (20 °C)
  • Flash Pt:173 °C
  • Storage Temperature:Ambient
  • EINECS:203-150-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to beige to grey to light brown Powder or crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    111 to 115 °C
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications