Acetanilide 99+%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: N-Phenylacetamide
- CAS Number:103-84-4
- Formula:C₈H₉NO
- MDL Number:MFCD00008674
- Molecular Weight:135.17 g/mol
- Merck Index:13,00051
- Boiling Pt:304 °C (760 mmHg)
- Melting Pt:111…115 °C
- Density:1,22 g/cm³ (20 °C)
- Flash Pt:173 °C
- Storage Temperature:Ambient
- EINECS:203-150-7
Documents
Specifications
Specification Test Results
- Appearance:White to beige to grey to light brown Powder or crystals or flakes
- Infrared spectrum:Conforms
- Melting point:111 to 115 °C
- GC:≥99.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



