Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl 4-hydroxybenzoate 99%

Ethyl 4-hydroxybenzoate 99%

Plastic bottle, 100 g
Catalog # ACRO150251000
Ethyl 4-hydroxybenzoate 99%
Catalog # ACRO150251000
CAS Number:  120-47-8
Ethyl 4-hydroxybenzoate  99%
Ethyl 4-hydroxybenzoate 99%

Plastic bottle, 100 g

CAS Number:  120-47-8
Price
26,20 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Merck Index:13,03869
  • Boiling Pt:297…298 °C (760 mmHg)
  • Melting Pt:115…118 °C
  • Density:1,078 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:204-399-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    115 to 118 °C
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications