Ethanolamine 99%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Aminoethanol, Monoethanolamine
- CAS Number:141-43-5
- Formula:NH₂CH₂CH₂OH
- MDL Number:MFCD00008183
- Molecular Weight:61.08 g/mol
- Boiling Pt:171 °C (1013 hPa)
- Melting Pt:10.3 °C
- Density:1.012 g/cm³
- Flash Pt:92 °C
- Storage Temperature:Ambient
- UN:2491
- ADR:8,III
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4530 to 1.4550 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
- Color scale:≤15 APHA
Specifications
- Pack type:Glass bottle
- Pk:1 L



