Acetaldehyde 99.5%, extra pure
Glass bottle, 10 mL
: Ethanal
- :75-07-0
- :CH₃CHO
- :MFCD00006991
- :44.05 g/mol
- :12,00037
- :21 °C (760 mmHg)
- :–123 °C
- :0.785 g/cm³
- :–27 °C
- :Refrigerator
- :200-836-8
- :1089
- :3,I
Documents
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3310 to 1.3330 (20°C, 589 nm)
- Water:≤0.02 %
- GC:≥99.4 %
- Chlorine (Cl):≤0.003 % (total)
- Pack type:Glass bottle
- Pk:10 mL



