Triiodomethane 99+%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Iodoform
- CAS Number:75-47-8
- Formula:CHI₃
- MDL Number:MFCD00001069
- Molecular Weight:393.73 g/mol
- Merck Index:13,05055
- Boiling Pt:218 °C (1013 hPa)
- Melting Pt:119…122 °C
- Density:4,1 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-874-5
Specifications
Specification Test Results
- Appearance:Bright yellow to dark yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:119 to 122 °C
- Loss on drying:≤0.5 % (24 h) (Over silicagel)
- Residue after ignition:≤0.1 %
- Titration:≥99.0 %
- Chloride (Cl):≤110 ppm
- Sulfate (SO4):≤170 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



