2,4-Dichlorophenol 99%
Glass bottle for solids, 100 g
: 1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene
- :120-83-2
- :C₆H₄Cl₂O
- :MFCD00002169
- :163 g/mol
- :13,03098
- :210 °C (1013 hPa)
- :41…44 °C
- :1,4 g/cm³ (20 °C)
- :113 °C
- :Ambient
- :204-429-6
- :2928
- :6.1,II
- Appearance:White to beige Crystalline solid
- Infrared spectrum:Conforms
- Melting point:41 to 44 °C
- Water:≤0.3 %
- GC:≥98.5 %
- Specific optical rotation:+9.0° to +11.0° (20 °C, 589 nm) (C=2 in methanol)
- Pack type:Glass bottle for solids
- Pk:100 g



