Order Entry
Export
Orders LinkContactUsLinkComponent
2,4-Dichlorophenol 99%

2,4-Dichlorophenol 99%

Glass bottle for solids, 5 g
Catalog # ACRO147720050
Toxic
Corrosive
Environment
2,4-Dichlorophenol 99%
Catalog # ACRO147720050
CAS Number:  120-83-2
2,4-Dichlorophenol 99%
2,4-Dichlorophenol 99%

Glass bottle for solids, 5 g

CAS Number:  120-83-2
Price
20,20 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene
  • CAS Number:120-83-2
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002169
  • Molecular Weight:163 g/mol
  • Merck Index:13,03098
  • Boiling Pt:210 °C (1013 hPa)
  • Melting Pt:41…44 °C
  • Density:1,4 g/cm³ (20 °C)
  • Flash Pt:113 °C
  • Storage Temperature:Ambient
  • EINECS:204-429-6
  • UN:2928
  • ADR:6.1,II

Documents

Specifications

Specification Test Results

  • Appearance:
    White to beige Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    41 to 44 °C
  • Water:
    ≤0.3 %
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +9.0° to +11.0° (20 °C, 589 nm) (C=2 in methanol)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications