Piperidine 99%, extra pure
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Piperidine 99%, extra pure
Catalog # ACRO147180025
Supplier: Thermo Fisher Scientific
CAS Number: 110-89-4
Jump to:
Product Details & Documents
Synonyms: Hexahydropyridine, Hexazane, Pentamethyleneimine
- CAS Number:110-89-4
- Formula:C₅H₁₁N
- MDL Number:MFCD00005979
- Molecular Weight:85.15 g/mol
- Merck Index:12,07621
- Boiling Pt:107.3 °C (1013 hPa)
- Melting Pt:–11 °C
- Density:0.862 g/cm³
- Flash Pt:16 °C
- Storage Temperature:Ambient
- EINECS:203-813-0
- UN:2401
- ADR:8,I
Documents
ACRO14718
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4510 to 1.4540 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥98.5 %
- Color scale:≤50 APHA
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



