Order Entry
Export
Orders LinkContactUsLinkComponent
 

D-(+)-Xylose 99+%

Plastic bottle, 500 g
  ACRO141005000
D-(+)-Xylose 99+%
  ACRO141005000
 :  58-86-6
D-(+)-Xylose 99+%
D-(+)-Xylose 99+%

Plastic bottle, 500 g

 :  58-86-6
 
238,00 €
   
1 * 500 g
 
  •  :58-86-6
  •  :C₅H₁₀O₅
  •  :MFCD00151475
  •  :150.13 g/mol
  •  :13,10142
  •  :331 °C (1013 hPa)
  •  :148…153 °C
  •  :1,525 g/cm³ (20 °C)
  •  :Ambient
  •  :200-400-7

 

 

 

  • Appearance:
    White to almost white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 % (K.F.)
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    +18.0° to +21.0° (20°C, 589 nm) (c=10, H2O/NH3)
  • Heavy metals (as Pb):
    ≤10 ppm

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g