D-(+)-Xylose 99+%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:58-86-6
- Formula:C₅H₁₀O₅
- MDL Number:MFCD00151475
- Molecular Weight:150.13 g/mol
- Merck Index:13,10142
- Boiling Pt:331 °C (1013 hPa)
- Melting Pt:148…153 °C
- Density:1,525 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-400-7
Documents
ACRO14100
SDS
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.2 % (K.F.)
- Sulfated ash:≤0.2 %
- HPLC:≥99.0 % (ELSD)
- Specific optical rotation:+18.0° to +21.0° (20°C, 589 nm) (c=10, H2O/NH3)
- Heavy metals (as Pb):≤10 ppm
Specifications
- Pack type:Plastic bottle
- Pk:100 g



