Urea 99.5% for analysis
Plastic drum, 5 kg
: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- :57-13-6
- :NH₂CONH₂
- :MFCD00008022
- :60.06 g/mol
- :13,09935
- :197 °C (1013 hPa)
- :131…135 °C
- :1,34 g/cm³ (20 °C)
- :Ambient
- :200-315-5
- :01-2119463277-33
- Appearance:White Crystals
- Infrared spectrum:Conforms
- Melting point:132 to 135 °C
- Residue after ignition:≤0.01 %
- Titration with HClO4:≥99.4 %
- Chloride (Cl):≤5 ppm
- Sulfate (SO4):≤10 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤10 ppm
- Insoluble matter:≤0.01 %
- Pack type:Plastic drum
- Pk:5 kg



