n-Undecane 99%
Glass bottle, 500 mL
: Undecane
- :1120-21-4
- :H₃C(CH₂)₉CH₃
- :MFCD00008959
- :156.31 g/mol
- :196 °C (760 mmHg)
- :–26 °C
- :0.74 g/cm³
- :65 °C
- :Ambient
- :214-300-6
- :2330
- :3,III
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4160 to 1.4180 (20°C, 589 nm)
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:500 mL



