Order Entry
Export
Orders LinkContactUsLinkComponent
3,3'-Thiodipropionic acid 99%

3,3'-Thiodipropionic acid 99%

Plastic bottle, 500 g
Catalog # ACRO138755000
Irritant
3,3'-Thiodipropionic acid 99%
Catalog # ACRO138755000
CAS Number:  111-17-1
3,3'-Thiodipropionic acid 99%
3,3'-Thiodipropionic acid 99%

Plastic bottle, 500 g

CAS Number:  111-17-1
Price
43,10 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  3,3-Thiodipropionic acid
  • CAS Number:111-17-1
  • Formula:C₆H₁₀O₄S
  • MDL Number:MFCD00002781
  • Molecular Weight:178.21 g/mol
  • Merck Index:12,09468
  • Boiling Pt:347 °C (1013 hPa)
  • Melting Pt:130…133 °C
  • Density:1,496 g/cm³ (20 °C)
  • Flash Pt:128 °C
  • Storage Temperature:Ambient
  • EINECS:203-841-3

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    130 to 133 °C
  • Titration with NaOH:
    98.5 to 101.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications