1,3-Propanediol 98%
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
1,3-Propanediol 98%
Catalog # ACRO131400010
Supplier: Thermo Fisher Scientific
CAS Number: 504-63-2
Jump to:
Product Details & Documents
Synonyms: 1,3-Propylene glycol, 1,3-Dihydroxy propane
- CAS Number:504-63-2
- Formula:HO(CH₂)₃OH
- MDL Number:MFCD00002949
- Molecular Weight:76.1 g/mol
- Merck Index:12,09845
- Boiling Pt:214 °C (760 mmHg)
- Melting Pt:–32 °C
- Density:1.052 g/cm³
- Flash Pt:140 °C
- Storage Temperature:Ambient
- EINECS:207-997-3
Specifications
Specification Test Results
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4380 to 1.4400 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥97.5 %
- Color scale:≤20 APHA
- UV:at 280 nm A: ≤0.301
- Residue after evaporation:≤0.001 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:ACRO131400010
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating)
- Environmentally Preferable:Safer Human and Environmental Health



