2-Phenylpropionic acid 98%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2-Phenylpropionic acid 98%
Catalog # ACRO130870250
Supplier: Thermo Fisher Scientific
CAS Number: 492-37-5
Jump to:
Product Details & Documents
Synonyms: (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
- CAS Number:492-37-5
- Formula:C₉H₁₀O₂
- MDL Number:MFCD00002650
- Molecular Weight:150.18 g/mol
- Boiling Pt:260…262 °C (760 mmHg)
- Melting Pt:5 °C
- Density:1.1 g/cm³
- Flash Pt:148 °C
- Storage Temperature:Ambient
- EINECS:207-752-0
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5210 to 1.5250 (20°C, 589 nm)
- GC:≥97.5 %
- Titration with NaOH:97.5 to 102.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



