Order Entry
Export
Orders LinkContactUsLinkComponent
2-Phenylpropionic acid 98%

2-Phenylpropionic acid 98%

Glass bottle for solids, 10 g
Catalog # ACRO130870100
Irritant
2-Phenylpropionic acid 98%
Catalog # ACRO130870100
CAS Number:  492-37-5
2-Phenylpropionic acid 98%
2-Phenylpropionic acid 98%

Glass bottle for solids, 10 g

CAS Number:  492-37-5
Price
57,30 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, DL-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid
  • CAS Number:492-37-5
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00002650
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:260…262 °C (760 mmHg)
  • Melting Pt:5 °C
  • Density:1.1 g/cm³
  • Flash Pt:148 °C
  • Storage Temperature:Ambient
  • EINECS:207-752-0

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5210 to 1.5250 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Titration with NaOH:
    97.5 to 102.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications