2-Phenylethanol 99%
Glass bottle, 5 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
- CAS Number:60-12-8
- Formula:C₆H₅CH₂CH₂OH
- MDL Number:MFCD00002886
- Molecular Weight:122.17 g/mol
- Merck Index:12,07370
- Boiling Pt:219 °C (760 mmHg)
- Melting Pt:–27 °C
- Density:1.02 g/cm³
- Flash Pt:102 °C
- Storage Temperature:Ambient
- EINECS:200-456-2
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5300 to 1.5350 (20°C, 589 nm)
- GC:≥98.5 %
- Specific gravity:(25/25°C) 1.016 to 1.021
- UV:at 220 nm A: ≤0.04
- Titration after Ion exchange:≥99.0 % (on dry substance)
- Membrane filtration test:(on 0.2 M soln.) Passes test
- Suitability:(HPLC gradient) Passes test
Specifications
- Pack type:Glass bottle
- Pk:5 mL



