Perylene 99+%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:198-55-0
- Formula:C₂₀H₁₂
- MDL Number:MFCD00004142
- Molecular Weight:252.32 g/mol
- Merck Index:12,07325
- Boiling Pt:443 °C (1013 hPa)
- Melting Pt:277…280 °C
- Density:1,362 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-900-9
Documents
ACRO13007
SDS
Specifications
Specification Test Results
- Appearance:Ochre to brown Powder
- Infrared spectrum:Conforms
- Melting point:277 to 280 °C
- HPLC:≥99.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



