2-Octanone 99+%
Glass bottle, 500 mL
: Octan-2-one
- :111-13-7
- :C₈H₁₆O
- :MFCD00009540
- :128.21 g/mol
- :13,04731
- :173 °C (760 mmHg)
- :–16 °C
- :0.819 g/cm³
- :55 °C
- :Ambient
- :203-837-1
- :1224
- :3,III
Documents
ACRO12942
SDS
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4140 to 1.4160 (20°C, 589 nm)
- GC:≥99.0 %
- Clarity of solution:Passes test
- Pack type:Glass bottle
- Pk:500 mL



