1,9-Nonanediol 99%
Glass bottle for solids, 100 g
: 1,9-Dihydroxy nonane, Nonane-1,9-diol
- :3937-56-2
- :C₉H₂₀O₂
- :MFCD00002991
- :160.26 g/mol
- :177 °C (15 mmHg)
- :44…48 °C
- :0.918 g/cm³
- :162 °C
- :Ambient
- :223-517-5
- Appearance:White Fused crystalline solid
- Infrared spectrum:Conforms
- Melting point:44 to 48 °C
- GC:≥97.5 %
- Pack type:Glass bottle for solids
- Pk:100 g



