1,9-Nonanediol 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,9-Dihydroxy nonane, Nonane-1,9-diol
- CAS Number:3937-56-2
- Formula:C₉H₂₀O₂
- MDL Number:MFCD00002991
- Molecular Weight:160.26 g/mol
- Boiling Pt:177 °C (15 mmHg)
- Melting Pt:44…48 °C
- Density:0.918 g/cm³
- Flash Pt:162 °C
- Storage Temperature:Ambient
- EINECS:223-517-5
Documents
ACRO12912
SDS
Specifications
Specification Test Results
- Appearance:White Fused crystalline solid
- Infrared spectrum:Conforms
- Melting point:44 to 48 °C
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



