Order Entry
Export
Orders LinkContactUsLinkComponent
2-Nitrobenzaldehyde 99+%

2-Nitrobenzaldehyde 99+%

Plastic bottle, 100 g
Catalog # ACRO128401000
Irritant
2-Nitrobenzaldehyde 99+%
Catalog # ACRO128401000
CAS Number:  552-89-6
2-Nitrobenzaldehyde 99+%
2-Nitrobenzaldehyde 99+%

Plastic bottle, 100 g

CAS Number:  552-89-6
Price
147,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:552-89-6
  • Formula:C₇H₅NO₃
  • MDL Number:MFCD00007132
  • Molecular Weight:151.12 g/mol
  • Merck Index:12,06684
  • Boiling Pt:153 °C (23 mmHg)
  • Melting Pt:41…44 °C
  • Density:1,35 g/cm³ (25 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:209-025-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow Powder or needles and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    41 to 45 °C
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications