Order Entry
Export
Orders LinkContactUsLinkComponent
1-Naphthol 99+%

1-Naphthol 99+%

Glass bottle for solids, 500 g
Catalog # ACRO128195000
Irritant
Corrosive
1-Naphthol 99+%
Catalog # ACRO128195000
CAS Number:  90-15-3
1-Naphthol 99+%
1-Naphthol 99+%

Glass bottle for solids, 500 g

CAS Number:  90-15-3
Price
71,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
  • CAS Number:90-15-3
  • Formula:C₁₀H₈O
  • MDL Number:MFCD00003930
  • Molecular Weight:144.17 g/mol
  • Merck Index:13,06409
  • Boiling Pt:278…280 °C (760 mmHg)
  • Melting Pt:95…97 °C
  • Density:1,224 g/cm³ (20 °C)
  • Flash Pt:125 °C
  • Storage Temperature:Ambient
  • EINECS:201-969-4
  • UN:2811
  • ADR:6.1,III

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    Beige to grey to brown Crystalline powder or crystals or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    95 to 97 °C
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications