S-Methylisothiouronium sulphate 98%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
S-Methylisothiouronium sulphate 98%
Catalog # ACRO127891000
Supplier: Thermo Fisher Scientific
CAS Number: 867-44-7
Jump to:
Product Details & Documents
Synonyms: S-Methylisothiourea sulphate, 2-Methyl-2-thiopseudourea sulphate, S-Methylthiouronium sulphate, 2-Methyl-2-thiopseudourea hemisulphate
- CAS Number:867-44-7
- Formula:C₄H₁₂N₄S₂·H₂SO₄
- MDL Number:MFCD00013055
- Molecular Weight:278.38 g/mol
- Melting Pt:240…241 °C (dec.)
- Storage Temperature:Ambient
- EINECS:212-759-7
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Titration with HClO4:≥97.5 %
- HPLC:≥97.5 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g



