Methylcyclohexane 99%, extra pure
Plastic drum, 10 L
Supplier: Thermo Fisher Scientific
Methylcyclohexane 99%, extra pure
Catalog # ACRO126580100
Supplier: Thermo Fisher Scientific
CAS Number: 108-87-2
Jump to:
Product Details & Documents
- CAS Number:108-87-2
- Formula:C₆H₁₁CH₃
- MDL Number:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–126.3 °C
- Density:0.77 g/cm³
- Flash Pt:–3 °C
- Storage Temperature:Ambient
- EINECS:203-624-3
- UN:2296
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4210 to 1.4240 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.01 %
Specifications
- Pack type:Plastic drum
- Pk:10 L



