2-Methylbutane 99+%, extra pure
Glass bottle, 1 L
: iso-Pentane
- :78-78-4
- :CH₃CH₂CH(CH₃)₂
- :MFCD00009338
- :72.15 g/mol
- :30 °C (760 mmHg)
- :–160 °C
- :0.62 g/cm³
- :–51 °C
- :Refrigerator
- :201-142-8
- :1265
- :3,I
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3530 to 1.3550 (20°C, 589 nm)
- GC:≥99.0 %
- Residue after evaporation:≤10 ppm
- Pack type:Glass bottle
- Pk:1 L



