Order Entry
Export
Orders LinkContactUsLinkComponent
2-Mercaptobenzothiazole 95%

2-Mercaptobenzothiazole 95%

Plastic bottle, 1 kg
Catalog # ACRO125450010
Irritant
Environment
2-Mercaptobenzothiazole 95%
Catalog # ACRO125450010
CAS Number:  149-30-4
2-Mercaptobenzothiazole 95%
2-Mercaptobenzothiazole 95%

Plastic bottle, 1 kg

CAS Number:  149-30-4
Price
64,50 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  2-Benzothiazolethiol, Mercapto-2 benzothiazole, Benzo[d]thiazole-2(3H)-thione, Benzothiazole-2-thiol, MBT
  • CAS Number:149-30-4
  • Formula:C₇H₅NS₂
  • MDL Number:MFCD00005781
  • Molecular Weight:167.26 g/mol
  • Merck Index:13,05893
  • Boiling Pt:302 °C (1013 hPa)
  • Melting Pt:172…182 °C
  • Density:1,42 g/cm³ (20 °C)
  • Flash Pt:243 °C
  • Storage Temperature:Ambient
  • EINECS:205-736-8
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    172 to 180 °C
  • Loss on drying:
    ≤0.5 % (60 °C, 3 hrs)
  • Sulfated ash:
    ≤0.5 %
  • Titration with NaOH:
    ≥94 %
  • Impurity:
    Contains 1 to 2% oil
  • UV:
    at 250 nm A: ≤0.05
  • Non-volatile matter:
    ≤0.0003 %
  • Acidity:
    ≤0.0002 meq/g
  • Alkalinity:
    ≤0.0002 meq/g

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications