Melamine 99%
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 2,4,6-Triamino-s-triazine, 1,3,5-Triazine-2,4,6-triamine, 2,4,6-Triamino-1,3,5-triazine
- CAS Number:108-78-1
- Formula:C₃H₆N₆
- MDL Number:MFCD00006055
- Molecular Weight:126.12 g/mol
- Merck Index:13,05834
- Melting Pt:354 °C (dec.)
- Density:1,57 g/cm³ (20 °C)
- Flash Pt:≥ 280 °C
- Storage Temperature:Ambient
- EINECS:203-615-4
Documents
ACRO12535
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



