Order Entry
Export
Orders LinkContactUsLinkComponent
 

(±)-Linalool 97%

Glass bottle, 100 mL
  ACRO125151000
 
(±)-Linalool 97%
  ACRO125151000
 :  78-70-6
(±)-Linalool 97%
(±)-Linalool 97%

Glass bottle, 100 mL

 :  78-70-6
 
37,30 €
   
1 * 100 mL
 
 :  (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
  •  :78-70-6
  •  :C₁₀H₁₈O
  •  :MFCD00008906
  •  :154.25 g/mol
  •  :13,05517
  •  :199 °C (760 mmHg)
  •  :0.861 g/cm³
  •  :75 °C
  •  :Ambient
  •  :201-134-4
 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4600 to 1.4640 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Specific gravity:
    (20/4°C) 0.859 to 0.864
  • Specific optical rotation:
    -2° to +2° (20°C, 589 nm) (neat)
  • Substance darkened by H2SO4:
    ≤10 APHA
  • Calcium (Ca):
    ≤0.5 ppm
  • Copper (Cu):
    ≤0.02 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.02 ppm
  • Magnesium (Mg):
    ≤0.05 ppm
  • Potassium (K):
    ≤0.1 ppm
  • Sodium (Na):
    ≤0.5 ppm
  • Zinc (Zn):
    ≤0.2 ppm
  • Total phosphorus (P):
    ≤0.1 ppm
  • Total silicon (Si):
    ≤0.05 ppm
  • Total sulfur (S):
    ≤0.001 %
  • Bromine number:
    ≤0.5

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL