Order Entry
Export
Orders LinkContactUsLinkComponent
Imidazole 99%

Imidazole 99%

Plastic bottle, 100 g
Catalog # ACRO122021000
Corrosive
Irritant
Health
Imidazole 99%
Catalog # ACRO122021000
CAS Number:  288-32-4
Imidazole 99%
Imidazole 99%

Plastic bottle, 100 g

CAS Number:  288-32-4
Price
43,80 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Glyoxaline, 1H-Imidazole, Glyoxalamine
  • CAS Number:288-32-4
  • Formula:C₃H₄N₂
  • MDL Number:MFCD00005183
  • Molecular Weight:68.08 g/mol
  • Merck Index:13,04935
  • Boiling Pt:255…256 °C (760 mmHg)
  • Melting Pt:86…90 °C
  • Density:1,18 g/cm³ (20 °C)
  • Flash Pt:145 °C
  • Storage Temperature:Ambient
  • EINECS:206-019-2
  • UN:3263
  • ADR:8,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Flakes
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications