2-Hydroxypyridine 97%
Plastic bottle, 50 g
: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
- :142-08-5
- :C₅H₅NO
- :MFCD00006268
- :95.10 g/mol
- :280…281 °C (760 mmHg)
- :102…109 °C
- :1,085 g/cm³ (30 °C)
- :Ambient
- :205-520-3
- :2811
- :6.1,III
- Appearance:Off-white to yellow to beige to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:103 to 108 °C
- HPLC:≥96.0 %
- Titration with HClO4:≥96.0 %
- Pack type:Plastic bottle
- Pk:50 g



