2-Hydroxypyridine 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
2-Hydroxypyridine 97%
Catalog # ACRO121840050
Supplier: Thermo Fisher Scientific
CAS Number: 142-08-5
Jump to:
Product Details & Documents
Synonyms: Pyridin-2-one, Pyridin-2(1H)-one, α-Hydroxypyridine, 2-Pyridone, α-Pyridone
- CAS Number:142-08-5
- Formula:C₅H₅NO
- MDL Number:MFCD00006268
- Molecular Weight:95.10 g/mol
- Boiling Pt:280…281 °C (760 mmHg)
- Melting Pt:102…109 °C
- Density:1,085 g/cm³ (30 °C)
- Storage Temperature:Ambient
- EINECS:205-520-3
- UN:2811
- ADR:6.1,III
Documents
ACRO12184
SDS
Specifications
Specification Test Results
- Appearance:Off-white to yellow to beige to brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:103 to 108 °C
- HPLC:≥96.0 %
- Titration with HClO4:≥96.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



