Order Entry
Export
Orders LinkContactUsLinkComponent
2'-Hydroxy-4'-methoxyacetophenone 99%

2'-Hydroxy-4'-methoxyacetophenone 99%

Glass bottle for solids, 10 g
Catalog # ACRO121330100
Irritant
2'-Hydroxy-4'-methoxyacetophenone 99%
Catalog # ACRO121330100
CAS Number:  552-41-0
2'-Hydroxy-4'-methoxyacetophenone 99%
2'-Hydroxy-4'-methoxyacetophenone 99%

Glass bottle for solids, 10 g

CAS Number:  552-41-0
Price
50,40 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  • CAS Number:552-41-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00008730
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:154 °C (20 -rr)
  • Melting Pt:47…50 °C
  • Density:1,1604 g/cm³ (40 °C)
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
  • EINECS:209-012-2

Documents

Specifications

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    47 to 50 °C
  • GC:
    ≥98.5 %
  • Stabilizer:
    Approx. 100 ppm Amylene
  • Residue after evaporation:
    ≤0.002 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications